Eisco Solar Cell with Wires
Supplier: Wards
PH201704
470356-240EA 14.65 USD
470356-240
Eisco Solar Cell with Wires
Educational Materials Physics Educational Materials Alternative Energy Learning Activities
Polycrystalline solar cell with wires used for educational purposes.
  • 2 V DC
  • 130 mA current
  • 5" cable length
Solar panel with two connected wires is used for educational study of solar and sustainable energy. Attached wires allow connection to multiple different pieces of equipment via soldering. Designed for use in a educational science setting including for use in various science projects. Can be used to power small motors or other loads.
Order Now
SPECIFICATIONS
Environmentally Preferable