AquaSprouts Fountains
Supplier: AQUASPROUTS, INC
6148 6155
470353-520EA 169.95 USD
470353-520 470353-524
AquaSprouts Fountains
Educational Materials Biology Educational Materials Ecology Learning Activities
Versatile indoor or outdoor system which can be used for Aquaponics, Hydroponics, or Water Garden.
  • Rated for indoor and outdoor use
  • Durable, weatherproof grow bed and fountain reservoir
  • Includes a submersible 160 gph pump
  • Control the flow rate with customizable plumbing fittings
  • Room for 6 or more plants
Add a touch of beauty and a splash of life to any room with the AquaSprouts Fountain! Water feature, planter, and eye-catching decor, the Fountain can be used as an aquaponic or hydroponic system, or a simple water garden.
Similar to AquaSproutsMore Product Information