Float and Spin Magnet Demonstrator
Supplier: United Scientific Supplies
FLSPMAG
470352-576EA 11 USD
470352-576
Float and Spin Magnet Demonstrator
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Demonstrate the wonders of magnetic levitation.
  • Smooth rotation
  • No batteries required
This attention-getting demonstration of magnetic repulsion uses a clear acrylic plate that keeps a spinning axle oriented above magnets built into the base of the apparatus. With a simple twist of your fingers, you can start this levitating magnet spinning and kick start a classroom discussion. The black and white coloring makes the demonstration easy to see and helps track the speed of the rotation. The axle measures 5” long.