Guardian 2000 Analog Hot Plates and Stirrers
Supplier: OHAUS CORP.
30680265 30680263 30680260 30680262 30680264 30680261 30680257 30680258 30680259
470350-994EA 328.5 USD
470350-994 470350-998 470350-986 470350-982 470350-996 470350-984 470350-988 470350-992 470350-990
Guardian 2000 Analog Hot Plates and Stirrers
Heaters Hot Plates
Rugged and reliable analog hotplates and stirrers for general purpose heating and stirring. They are ideal for lab environments that require affordable, everyday fast heating and reliable stirring. All three sizes offer simple knob control to easily adjust the heating and stirring functions.
  • Powerful stirring and fast heating at an economical price
  • Safely heat up to 500 °C and stir up to 15 L with easy to use knob control for simple and reliable everyday heating and stirring in the lab
Designed to last: The rugged metal housing has a spill resistantMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable