SureAir™ PCR Workstation
Supplier: Ward's
B5200 B5200-E B5200-FIL B5200-PRE
470350-922EA 3159.4 USD
470350-922 470350-924 470350-928 470350-926
SureAir™ PCR Workstation
Fume Hoods and Enclosures PCR Workstation Enclosures
The SureAir™ is designed to keep your workspace clean, easy-to-use and organized.
  • HEPA filter and UV light reduce contamination
  • Highly compact, 22×16" footprint
  • Includes cable access port - faciliates use of centrifuges, vortexers and other instruments
  • Integral pipette holder and shelf provide storage for pipettes and tips
  • Provides ISO 5 level clean conditions
The Benchmark SureAir™ PCR workstation is a compact, convenient way to reduce contamination concerns and provide a clean room environment at a fraction of the space and cost. The high-poweredMore Product Information