Solar Cells, Monocrystalline
Supplier: NEWARK ELECTRONICS
94W9302 94W9305
470350-852EA 20.55 USD
470350-852 470350-856
Solar Cells, Monocrystalline
Educational Materials Physics Educational Materials Alternative Energy Learning Activities
Maximum efficiency monocrystalline solar cells.
  • Encapsulated with low reflecting water clear epoxy to protect against physical impact and harsh outdoor environments
  • Terminated with a DC barrel plug Calculating Watts Power [Watt] = Voltage [Volt] Current [Ampere]
For applications requiring a higher voltage supply source, you can wire multiple solar panels in a series. Wire the positive terminal of solar panel #1 to the load, and connecting the negative terminal of solar panel #1 to the positive terminal of solar panel #2. In this wiring, the totalMore Product Information