Cardboard Spectrometer Kit
Supplier: Science First
654-0025
470350-112PK 121.29 USD
470350-112 470350-102
Cardboard Spectrometer Kit
Educational Materials Earth Science Educational Materials Astronomy Learning Activities
Observe absorption lines in the Sun's spectrum, the spectra of energized elements in gaseous states and the double yellow lines of Mercury vapors in the spectrum of a fluorescent lamp.
  • Available in multiple pack sizes
  • Recommended for astronomy, chemistry, physics, physical science and earth science at the high school through college level
The ready-to-assemble spectrometer comes with a strip of film and a scale from which a color may be identified by wavelength in nanometers or photon energy in electron volts.