Life/form® Basic Sanitary CPR Dog
Supplier: Nasco
LF01155 LF01156 LF01157 LF01158
470348-734EA 2617 USD
470348-734 470348-730 470348-684 470348-690
Life/form® Basic Sanitary CPR Dog
Models Zoology Models
Learn canine CPR.
  • Basic and advanced options
  • Muzzles can be disinfected
  • 5-year warranty
The Life/form® basic sanitary CPR dog provides each student with their own sanitary muzzle and nosepiece with a one-way valve. The muzzles may be disinfected and reused, while the nosepieces and lower airways are disposable.
In addition to practice in assisted breathing, students may practice the appropriate rate, pressure, and position of cardiac massage and coordinate the respiratory cardiovascular functions. A visible chest rise appears when correctMore Product Information