Nasco Healthcare® Breast Self Examination Trainer
Supplier: Nasco
160-1040
470348-732EA 387.55 USD
470348-732
Nasco Healthcare® Breast Self Examination Trainer
Models Human Biology Models
Realistic and easy to use.
  • Five different lumps
  • Gel material simulates a real feel
This Breast Self Examination (BSE) Trainer is a flesh based product for realistic practice. You will no longer have to struggle with keeping the exam model clean because the new, non toxic material is very stain resistant and cleans up easily with mild soap. Within the breast resides five lumps representing different disease states: cysts (fluid related), fibroadenomas (calcifications), sclerosing adenosis, or fat necrosis.
The filling is a water based gel thatMore Product Information