Eisco Optics Bench and Accessories, 2 m
Supplier: Wards
PH06502M
470346-454EA 457.8 USD
470346-454
Eisco Optics Bench and Accessories, 2 m
Educational Materials Physics Educational Materials Optics Learning Activities
Extra-long optics bench with multiple accessories included at an affordable price.
  • Robust rider profile, made from solid aluminum
  • White fillets for easy reading even in the dark
  • Light source and multiple accessories included
This premium optics bench is practical and smart. Made from aluminum, with adjustable leveling feet, it measures a full two meters long. Double graduations, and white fillets on both sides of the riders, makes it easy to make observations from either side of the bench, even in the dark. An included LED light source and 12 V powerMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable