Eisco Glass Graduated Cylinders, Squat Form
Supplier: Wards
CH0343M CH0343K CH0343J CH0343N CH0343L CH0343O CH0343I
470343-632EA 31.4 USD
470343-632 470344-008 470344-046 470344-858 470345-452 470345-844 470346-124
Eisco Glass Graduated Cylinders, Squat Form
Cylinders Measuring Cylinders
Specially designed to take up half the height of similar capacity tall-form cylinders.
  • Made of high quality, heavy duty, borosilicate 3.3 glass
  • Class A tolerance conforms with DIN ISO 4788 standards for accuracy
  • Spout for easy pouring
  • Hexagonal base provides maximum stability and prevents rolling when placed on side
  • Easy-to-read blue graduations and specifications contrast against light color liquids
Autoclavable, with superior chemical durability. Low temperature gradient - can withstand high temperatures and thermal shocks. Calibrated to contain.