Eisco 3-Piece Large Glass Erlenmeyer Flask Set
Supplier: Wards
CH0424LGSET
470345-464EA 58.4 USD
470345-464
Eisco 3-Piece Large Glass Erlenmeyer Flask Set
Flasks Erlenmeyer Flasks
Erlenmeyer flask set consisting of three larger sizes: 500, 1000 and 2000 ml, all made from borosilicate glass with narrow necks, white markings, and beaded rims.
  • Set includes one each 500, 1000 and 2000 ml Erlenmeyer flasks
  • Made of high quality, heavy duty, borosilicate 3.3 glass for excellent durability and chemical resistance
  • Narrow neck and conical shape prevents spills
  • Chip and crack resistant, fire polished beaded rims
  • Easy-to-read white enamel graduations and specifications for contrast against dark liquids
This 3-piece narrow neck ErlenmeyerMore Product Information