Eisco Meker Burner
Supplier: Wards
CH0098E CH0098F
470344-314EA 92.35 USD
470344-314 470345-426
Eisco Meker Burner
Heaters Gas Burners
Produces a shorter and hotter flame than standard bunsen burners.
  • Premium quality Meker burner that produces a short and very hot flame
  • Suitable for use with LPG (liquid petroleum gas) or butane gas
  • Serrated inlet arm accepts 6 to 8 mm tubing
  • Burner is mounted to a highly stable octagonal base
  • Both air and gas supply can be adjusted, allowing for separate control of flame height and temperature. Gas control is managed easily with a thumb knob on the bottom of the burner and air supply is controlled by adjusting the air ports
The Meker burner producesMore Product Information