Eisco Glass Graduated Cylinders with Hexagonal Base
Supplier: Wards
CH0345I CH0345N CH0345C CH0345H CH0345M CH0345F CH0345K CH0345JWT CH0345J CH0345B CH0345D CH0345BWT CH0345DWT CH0345A10 CH0345A CH0345FWT CH0345L CH0345G CH0345O CH0345CWT CH0345E
470343-644EA 11.65 USD
470343-644 470343-750 470343-828 470343-918 470344-050 470344-094 470344-324 470344-604 470344-756 470344-790 470344-814 470344-878 470345-030 470345-152 470345-256 470345-266 470345-380 470345-442 470345-696 470345-762 470345-840
Eisco Glass Graduated Cylinders with Hexagonal Base
Cylinders Measuring Cylinders
High-quality, hexagonal-based borosilicate cylinders with class A and class B tolerances conform with DIN ISO standards for accuracy.
  • Made of high quality, heavy duty, borosilicate 3.3 glass
  • Class A and class B tolerances conform with DIN ISO 4788 standards for accuracy
  • Hexagonal base provides maximum stability and prevents rolling when placed on side
  • Easy-to-read graduations and specifications
Spout for easy pouring, and hexagonal base. Autoclavable, with superior chemical durability. Low temperature gradient can withstand high temperatures andMore Product Information