Eisco® Hooked Stainless Steel Weights, Set of 9
Supplier: Wards
PH0037DSS
470344-170EA 160.85 USD
470344-170
Eisco® Hooked Stainless Steel Weights, Set of 9
Weights Calibration Weights
Stainless steel hooked weights set with a capacity of 1000 g. Ideal for classroom or lab experiments.
  • Stainless steel weights
  • Premium wooden block
  • Weights have hooks above and below main body, but can sit flat on a surface due to retractable bottom hook
  • Will last for decades of lab and classroom use
Eisco Labs set of nine steel hooked weights, 10 to 1000 g in wooden block. Precisely machined hooked weights. Set contains (1) 1000 g, (1) 500 g, (2) 200 g, (1) 100 g, (1) 50 g, (2) 20 g, (1) 10 g.