Eisco Polypropylene Petri Dishes with Lid
Supplier: Wards
CH0372BPK12 CH0372DPK6 CH0372EPK6 CH0372APK12 CH0372CPK12
470343-762EA 12.55 USD
470343-762 470344-200 470344-428 470345-550 470345-646
Eisco Polypropylene Petri Dishes with Lid
Culture Dishes Petri Dishes
Reusable Petri dishes with lid, molded in translucent polypropylene.
  • Non-toxic, non-sterile, multifunctional and ready to use
  • Made of high quality, unbreakable polypropylene that features high heat tolerance, and good to excellent chemical resistance
  • Translucent for easy examination of culture growth
  • Reusable and simple to clean. Top rack dishwasher safe
Excellent for general laboratory applications, including cell culturing, plant germination, precipitate drying and solvent evaporation, home or classroom science experiments, small storage solutions and more.