Biomembrane Model
Supplier: American 3B Scientific
1024034
470343-256EA 383.35 USD
470343-256
Biomembrane Model
Models Molecular Biology Models
Interactive biomembrane model.
  • Shipped unassembled
  • Learn about membrane structure
  • Includes materials for membrane protein simulation
Model to demonstrate the structure and function of a biomembrane. The structure of a lipid bilayer with various integrated and attached proteins is shown. By using liquids, the fluidity of the membrane can be impressively demonstrated. The membrane proteins can be moved with the supplied magnet.
Use this model as a teaching tool when discussing topics related to the cell membrane such as plasma membrane structure andMore Product Information