Foam Calorimeters
Supplier: United Scientific Supplies
UNFMCL01-PK/10
470340-282EA 40.9 USD
470340-282
Foam Calorimeters
Calorimeters
Economical foam calorimeter to investigate heat transfer.
  • Lids hold thermometers securely
  • Foam helps reuce thermal energy lost to the air
  • Exterior dimensions: 4.25"H×4"dia.
  • Interior dimensions: 4"H ×4"dia.
These foam calorimeter cups have excellent insulating properties so users can investigate thermal energy with minimal loss to the environment. The lids have holes for inserting a thermometer or temperature probe.
The maximum water capacity with lid on is 400 ml.
Order Now
SPECIFICATIONS
Environmentally Preferable