Eco- Mini Centrifuge
Supplier: United Scientific Supplies
UNMCNTF-4
470338-936EA 208.1 USD
470338-936
Eco- Mini Centrifuge
Centrifuges Microcentrifuges
A small centrifuge with lots of power.
  • Spins at 4000 rpm
  • Compact design
  • Low noise and vibration levels
This benchtop microcentrifuge is ideal for quick spin-downs of microcentrifuge tubes. Includes a multi-rotor that accommodates twelve microcentrifuge tubes in 0.2, 0.5, 1.5, or 2 ml capacities, or four 8-well, 0.2 ml PCR strips. Safety lid with lock stops centrifuge quickly when opened. Includes power adapter and user manual.
Technical specification:
Multi-rotor: holds 12 microcentrifuge tubes (0.2, 0.5, 1.5, or 2 ml) or 4 PCR strips (8-well, 0.2More Product Information