Snap Cap Container Vials, Polystyrene, Dynalon
Supplier: Dynalon
426364-13
470331-654CS 220.8 USD
470331-654
Snap Cap Container Vials, Polystyrene, Dynalon
Vials
Plastic snap cap vials are molded from rigid-wall, FDA compliant styrene with tight fitting polyethylene snap-on caps. Plastic snap cap container vials are excellent for storing dry materials, powders, tablets and more.
  • FDA compliant material
  • Transparent finish
There are approximately 16 drams per fluid ounce. The 7 dram size may be used with automatic cell counters for collection of sample and, where venous samples are used, for dilution purposes. The maximum operating temperature for polystyrene snap cap vials is 70 °C. The LDPE snap caps willMore Product Information