Using CRISPR to treat Cystic Fibrosis
Supplier: Edvotek
135
470330-674EA 134 USD
470330-674
Using CRISPR to treat Cystic Fibrosis
Educational Materials Biology Educational Materials Genetics Learning Activities
Hands-on simulation of CRISPR-Cas9 to target and treat Cystic Fibrosis.
  • Teach your students about new CRISPR technology
  • Includes all required reagents
  • Use DNA electrophoresis to view reach a diagnosis.
  • Activity can be completed in 45 minutes
In this experiment, students will simulate the use of CRISPR-Cas9 to target a genetic mutation found in a patient suffering from Cystic Fibrosis.
Students will develop an understanding of guide RNA (gRNA) design, and use agarose gel electrophoresis to examine pre-prepared DNA samples after CRISPR treatment.