VWR® Mini Magnetic Stirrers
Supplier: VWR International
470329-998
470329-998EA 241.3 USD
470329-998
VWR® Mini Magnetic Stirrers
Stirrers Magnetic Hotplate Stirrers
These rugged, compact and reliable units are ideal for educational laboratories and can stir up to 1L of water.
  • 5×5" square aluminum top plate
  • Stirring speed range 100 to 1200 rpm
  • 1000 ml capacity
Durable cast aluminum top plate will not chip or crack. A built-in support rod holder with locking knob accepts an optional support rod and clamp kit. Optional support plate allows the stirrer to be mounted to lab frames or ring stands.
Corresponding magnetic hotplate stirrers (heating and stirring 470329-996 and 470330-712) or hotplates (heating only 4703More Product Information