Spatulal and Spoon Tools
Supplier: GSC International
4-18351-PK
470313-332EA 10.9 USD
470313-332
Spatulal and Spoon Tools
Spatulas
This nickel-plated spoon and spatula tool is 14.9 cm long.
The bowl of its spoon measures 30 mm by 22 mm and has a 2 ml capacity, while the spatula measures 35 mm by 18 mm.
Order Now
SPECIFICATIONS
Environmentally Preferable