Nitric Acid

Supplier: ALDON CORP SE

NN0277-1L NN0280-500ML NN0276-2.5LR NN0276-500MLR NN0275-500ML NN0275-1L NN0269-500ML
470045-594EA 14.25 USD
470045-594 470301-537 470301-539 470301-540 470301-886 470301-884 470301-888
Nitric Acid
Chemicals for Science Education

CAS Number: 7697-37-2
Formula: HNO3
Density: 1.37—1.42 g/mL
Boiling and Freezing Point: 120 to 122 °C, -22 to -41°C
Solubility: Miscible with Water
Synonyms: Azotic Acid
Shelf Life: 12 Months


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.


Caution: This product is a corrosive and toxic oxidizer.

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR