Amylase

Supplier: ALDON CORP SE

AA0358-25G
470300-272EA 24.05 USD
470300-272
Amylase
Chemicals for Science Education

CAS Number: Mixture
Boiling Point: Decomposes
Synonyms: Bacterial Amylase 50,000 BAU/g, Alpha-Amylase
Shelf Life: 36 Months
Storage: Green


Note: The chemical specifications for some items on this product page may differ from the specifications listed above. For more information, consult the Safety Data Sheet or contact your Ward's representative.


Delivery information: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR