Arduino Leonardo Development Board
Supplier: NEWARK ELECTRONICS
07W3935
470237-742EA 32.19 USD
470237-742
Arduino Leonardo Development Board
Educational Materials Physics Educational Materials Applied Physics Learning Activities
Based on the ATmega32u4.
  • Used for sensing and instrumentation
  • Perfect for advanced programming use
  • ATmega32u4 8bit AVR RISC based microcontroller
  • Arduino Leonardo can be powered via the micro USB connection or with an external power supply
  • The board can operate on an external supply of 6 to 20 V
  • Built in LED connected to digital pin 13; When the pin is high value, the LED is on
  • The board appears as a generic keyboard and mouse, and can be prog to control these input devices
  • USB Over current protection
This board has 20 digital input/output pins (ofMore Product Information