Innovation Biological Microscopes, LW Scientific
Supplier: LW Scientific
INM-T05A-IPL3 INM-B05A-IPL3 INM-T04A-IPL3 INM-B04A-IPL3
470231-946EA 3301.05 USD
470231-946 470231-948 470231-950 470231-952 470231-954
Innovation Biological Microscopes, LW Scientific
Microscopes
Premium choice for discerning cytologists,doctors, and researchers.
  • The eyepieces are the largest and widest-viewing eyepieces in this microscope class
  • The siedentopf-type head is inclined 30-degrees for a comfortable user posture
  • The rackless, belt-drive system allows the stage to operate smoothly, comfortably, and safely
  • Its ergonomic, durable design ensures comfort and dependability for hours of daily use
  • The bright, white, uniform LED illumination maintains constant daylight-color, showing stained specimens in their true vibrant colors