Hermle Universal Benchtop Centrifuges
Supplier: Ward's
Z326 Z326-K Z306-100 Z326-100H Z326-100H-AC Z326-02MP Z326-2420 Z326-2420H Z326-3020H Z326-4420 Z326-6405 Z326-3202 Z326-1205 Z326-1215 Z326-3015 Z326-0650-LS Z326-0685-25 Z326-0650-HS Z326-1050 Z326-2010 Z326-BND Z326-K-BND
470230-644EA 7556 USD
470230-644 470230-646 470230-648 470230-650 470230-652 470230-654 470230-656 470230-658 470230-660 470230-662 470230-664 470230-666 470230-668 470230-670 470230-672 470230-674 470230-676 470230-678 470230-680 470230-682 470230-684 470336-498 470336-500
Hermle Universal Benchtop Centrifuges
Centrifuges Benchtop Centrifuges
Setting new standards in this centrifuge class.
  • Bundles are available with our most popular rotor choices
  • Models available with or without refrigeration
  • 19 different rotor choices available to choose from
  • Suitable for nearly every laboratory application
With 19 available rotor options, the Z326 defines the term “universal”. Swinging bucket rotors and high speed angled rotors are available to accept tubes from 0.2 ml up to 85 ml, making this centrifuge suitable for nearly any laboratory application. Despite an impressive capacity of up to 4×100 ml (or 6More Product Information