Light Emitting Diodes
Supplier: Wards
PH1019A
470230-382PK 7.1 USD
470230-382
Light Emitting Diodes
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Diodes for PCB or panel mount.
  • Red LED 5 mm standard high quality with diffused lens
  • Maximum voltage 3.0 V D.C. PCB or panel mount
  • Pack of 10
Labs Red LED 5 mm standard high quality with diffused lens. Maximum voltage 3.0 V D.C. PCB or panel mount. Pack of 10