Lenz’ Law
Supplier: Wards
PH0802LLO
470230-270EA 25.35 USD
470230-270
Lenz’ Law
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Loop apparatus ideal for studying Lenz Law and Faraday's Law.
  • Blue cast iron base with rod and needle for holding loops
  • Open loop dimensions: Inner dia. 1.82"/Outer dia. 1.98"
  • Closed loop dimensions: Inner dia. 1.82"/Outer dia. 1.98"
  • Aluminum loops
  • Loops connected by straight bar with groove for placement on needle
The Lenz' Law apparatus with both open and closed loops is effective for studying and demonstrating both Lenz' Law and Faraday's Law.