Pendulum Clamp Aluminum
Supplier: Wards
PH0308
470230-114EA 32.45 USD
470230-114
Pendulum Clamp Aluminum
Educational Materials Physics Educational Materials Mechanics Learning Activities
Aluminum pendulum clamp with screw clamps for attaching to rods.
  • For use in demonstrations or experiments involving pendulums
  • Can attach up to three pendulums to three screw clamps
  • Clamp attaches to support rods up to 16 mm in dia.
This sturdy pendulum clamp is constructed of an aluminum support bar with three screw clamps, attached to a die cast head clamp for use on support rods to hang weights and pendulums. Can attach to support rods up to 16mm in diameter. Three screw clamps can securely and individually be used to attach weights with strings.More Product Information