Phlebotomy Tray
Supplier: United Product & Instrument
48700
470229-850EA 102.5 USD
470229-850
Phlebotomy Tray
Trays/Dishes/Pots Blood Collection Trays
Have Everything At Hand.
  • Constructed of tough LDPE
  • Durable and easy to clean
  • Includes Tube-Cube™
A simple and compact design for the busy phlebotomist. The Flexi-Phleb has large carrying capacity without bulk. The tray provides a handy pull out drawer that is great for storing needles, bandages and other items. The top level accepts removable Tube-CUBE™ racks that secure a variety of tubes, ice cups for temperature sensitive samples, sharps containers, or whatever your specific activities demand. Back at the lab, merely exchange your collected samplesMore Product Information