Red Planet Telescope
Supplier: CARSON OPTICAL
RP-200
470227-892EA 287 USD
470227-892
Red Planet Telescope
Telescopes
A Refractor telescope.
  • 80 mm Aperture
  • 500 mm Focal Length
The telescope features superior, fully coated optics, an 8 x 21 mm finder scope, two high-quality eyepieces and a 45° corner lens. The eyepieces are K 20 mm and K 9 mm. The corner lens flips the image right-side up; great for terrestrial and astronomical viewing.
The Red Planet features a heavy-duty aluminum tripod and Horizon mount with slow motion cables to track celestial objects easily.