3B Scientific® Meiosis Model
Supplier: American 3B Scientific
1013869
470224-958EA 807.3 USD
470224-958
3B Scientific® Meiosis Model
Models Molecular Biology Models
The three-dimensional relief model shows the 10 stages of meiosis on the basis of a typical mammal cell.
  • Chromosomes colored according to modified AZAN staining colors
  • Cell components are color-coded in accordance with educational aspects
  • Attaching magnets on the rear
  • Storage system, free-standing or hanging up
  • Supplied with detailed description and copying templates
  • 3 year manufacturers warranty
1. Interphase (stage of G1-phase), 2. Prophase I (leptotene), 3. Prophase I (zygotene and pachytene), 4. Prophase I (diplotene), 5. Prophase I (diakinesis), 6More Product Information