Green Fuel Cell Lab Activity
Supplier: Ward's Science
IS6500
470221-558EA 84.5 USD
470221-558
Green Fuel Cell Lab Activity
Educational Materials Environmental Educational Materials Environmental Learning Activities
The Green Fuel Cell Lab Activity is a great introduction to green energy production.
  • Users Monitor the Voltage Generated by a Biological Fuel Cell
  • Ideal Introduction to Green Energy Production
  • Methylene Blue Works as an Electron Mediator
Create a biological fuel cell using the yeast Saccharomyces cerevisiae. A simple sugar is used as a food source for the yeast, and methylene blue is employed as an electron mediator. The cell is constructed without the mediator, and a baseline voltage is established as some electrochemical activity occurs.