pH Indicator Papers
Supplier: Precision Laboratories
190-12V-100
470024-816CS 19.37 USD
470024-816 470024-850 470151-048 470150-838 470220-896
pH Indicator Papers
Test Paper pH Indicator Papers and Test Strips
pH Test Papers in your Choice of Two Indicator Colors.
  • Easy to read
  • Vial of 100
Litmus red paper changes to blue at pH 8.1. Color comparison chart not included.