Density ID Set

Supplier: Wards
PH0113I
470220-778EA 70.05 USD
470220-778
Density ID Set
Educational Materials Physics Educational Materials Properties of Matter Learning Activities in Physics
The Density ID set offers hands-on learning on the concept of density.

  • Set of 12 Cylindrical Bars of Different Length and Material
  • Wooden Holder Included
  • Materials Include Softwood, Rubber, Pine, and More
  • Students Learn that a Substance's Density is Constant Even as Its Mass and Volume Change

The set includes 12 cylindrical bars of different lengths and materials. A wooden holder stores the set.

Ordering information: This set includes hardwood (oak), softwood (pine), aluminum, copper, brass, rubber, nylon, plc., Delrin (polyoxymethylene), glass, Lucite, and Teflon.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR