Eisco Gold Leaves for Electroscope
Supplier: Wards
PH0914
470219-524PK 24.35 USD
470219-524
Eisco Gold Leaves for Electroscope
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Enhance your electroscope with these fine gold leaves.
  • Pack of 6 gold leaves
  • Replacements for gold leaves in electroscope
  • Leaf Dimensions: 1.25" long and 0.5" wide
  • Provided in plastic case
The gold leaves are spares for use in an electroscope in the event that one misplaces or damages the gold leaves used with the device (pack of 6 gold leaves). The electroscope is a tool that is used to study basic electrostatic principles.