PYREX® Crystallizing Dishes
Supplier: Corning
3140-100
470211-452CS 587.6 USD
470211-452
PYREX® Crystallizing Dishes
Trays/Dishes/Pots Crystallizing Dishes
PYREX® borosilicate glass, without spout.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Flat bottom and straight sides. Rims reinforced and fire-polished to reduce chipping.