VWR® Traceable® Spatula Balance
Supplier: VWR International
470204-724
470204-724EA 84.15 USD
470204-724
VWR® Traceable® Spatula Balance
Balances and Scales
The intelligent function and intuitive operation of this unique balance add a degree of convenience and professionalism to any lab.
  • Intelligent functionality
  • Intuitive operation
  • Detachable scoop
  • Digital display on comfortable handle
The spatula simultaneously scoops material and instantly displays its weight. A digital display is located directly on the comfort-grip handle and the detachable scoop features graduated markings for accurate measurements. Additional features include a hold function, tare button, and modes of both grams and ounces.