Promega® PCR Master Mix (2x)*
Supplier: PROMEGA SUNNYVALE INC SE
M7502
470203-876EA 183.65 USD
470203-876
Promega® PCR Master Mix (2x)*
Nucleic Acid Reagents End-point PCR Enzymes and Kits
The premixed ready-to-use solution contains Taq DNA polymerase, dNTPs, and magnesium chloride at optimal concentrations for efficient amplification of DNA templates by PCR. Enough for 100 reactions (2.5 mL).
Note: Can be stored at 4°C for up to six weeks or -20° C for longer.
The premixed ready-to-use solution contains Taq DNA polymerase, dNTPs, and magnesium chloride at optimal concentrations for efficient amplification of DNA templates by PCR
Enough for 100 reactions (2.5 mL)