Electronics Snap Circuits Kits
Supplier: Elenco Electronics
SC-100-A SC-100
470007-248KT 58.8 USD
470007-248 470111-126 470200-470
Electronics Snap Circuits Kits
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Electronics Snap Circuit Kits feature easy-to-connect components, making them perfect for elementary electronics labs.
  • Easy-To-Connect Components
  • Instructions Included
  • Color-Coded Parts
  • Designed for Students New to Circuit Assembly
  • Projects Include Switch Circuits through Motors, Logic Circuits, and More
The circuit components are firmly attached to plastic bases, color coded by function, and mounted on snap bases for easy assembly and reliable contact.
When laid out on the base, the functioning circuit resembles the conventional circuit diagram,More Product Information