SP Bel-Art Easy-Read Environmentally Friendly, General Purpose Liquid-In-Glass Thermometers, Bel-Art Products, a part of SP
Supplier: Bel-Art Products
B60304-0200
470200-276EA 31.3 USD
470200-276
SP Bel-Art Easy-Read Environmentally Friendly, General Purpose Liquid-In-Glass Thermometers, Bel-Art Products, a part of SP
Thermometers Analog Thermometers
EnviroKleen™ certified thermometers feature black, non-toxic, biodegradable, Enviro-Safe® liquid.
  • Black liquid against lead-free, yellow back glass for easy readability and fewer reading errors
  • Manufactured and tested by our USA quality team
Ideal for use in chemical/petrochemical laboratories, universities, scientific institutions, food and beverage processing, pharmaceutical and medical industries, dairies, breweries and distilleries.
Certifications: Traceable to NIST.
Ordering information: Includes multi-language (English, French, German,More Product Information