Iodamoeba butschlii, Trophozoites and Cysts
Supplier: Avantor
470181-866EA 7.75 USD
470181-866
Iodamoeba butschlii, Trophozoites and Cysts
Slides Prepared Slides
Non-pathogenic Amoeba.
  • Identifying features clearly distinguishable
  • Rigorous quality control standards
Commensal organism living in the large intestine of humans. It possesses a large, commonly off-center karyosome.