Saccharomyces cereviciae Strains for Genetic Manipulation
Supplier: Ward's Science
CALHAO CALHBO CALHBRT CALHBT CALADE2 CALHART CALHAT CALA2ADE
470177-266EA 13.95 USD
470177-266 470177-268 470177-270 470179-678 470179-680 470179-682 470179-684 470179-686
Saccharomyces cereviciae Strains for Genetic Manipulation
Specimens Live Specimens Fungi - Living Material
Yeast strains are ideal for genetic study because they are easy to grow and maintain, their life cycle is complete in approximately one week, and you can arrest different stages of the life cycle by growing it on different types of media.
  • Easy to grow and maintain
  • Variety of strains available
  • All strains are haploid form
  • Supplied on YEAD agar, incubate at 30°C
We offer a variety of strains that carry the traits for color and tryptophan (Trp) and adenine (Ade) dependence or independence, expressed as either different color colonies or by their abilityMore Product Information