Ward's® ATP Muscle Kit
Supplier: JEFFREY M BUBEL
380025 RR0001-KIT
470176-820KT 74.9 USD
470176-820 470176-822 470177-242
Ward's® ATP Muscle Kit
Educational Classroom Kits and Activities
Discover How Muscles Contract
  • Purchase the complete kit only once, then get refills as you need them.
Strips of psoas muscle are available in a tube of 50% glycerol for study of ATP induced contractions and muscle striation. Each strip provides enough material for four to eight students. The solution must be stored at -4 °C until you are ready to use it.
Full Kits allow students to perform several demonstrations, including identification of muscle microanatomy, stimulation of muscle contraction with ATP solution, and correlation of chemicals to theMore Product Information