Polypropylene Buchner Funnels
Supplier: United Scientific Supplies
57105
470174-672EA 16.4 USD
470174-672
Polypropylene Buchner Funnels
Funnels Filtration Funnels Büchner Funnels
Unbreakable, polypropylene funnels are ideal for classroom use. Sturdy two piece construction with tight fitting bottom cup and stem permits vacuum filtration. Can be autoclaved.
Order Now
SPECIFICATIONS
Environmentally Preferable