Go Direct™ Spectrovis Plus
Supplier: Vernier Software & Technology
GDX-SVISPL
470166-544EA 478.99 USD
470166-544
Go Direct™ Spectrovis Plus
Spectrophotometers and Photometers UV/Visible Spectrophotometers
Quickly measures a full wavelength spectrum.
  • Determine peak wavelength to collect data on solution concentration for studies of Beer’s law or to monitor rates of reaction
  • Collect a full wavelength spectrum to measure absorbance, percent transmittance, fluorescence (at 405 nm or 500 nm excitation), or emissions
  • Conduct enzyme kinetics experiments
  • Engage in equilibrium studies of absorbance vs. time or absorbance vs. concentration
Introduce your students to spectroscopy with the affordable Go Direct SpectroVis Plus Spectrophotometer. Capable ofMore Product Information