Ward's® Comprehensive Periodic Table
Supplier: MERCURY PRINT PRODUCTIONS SE
1600 1000_U 1300_U 1400_U
470165-562CS 585.09 USD
470165-562 470165-570 470217-482 470217-486 470217-488
Ward's® Comprehensive Periodic Table
Charts
This chart displays detailed and up-to-date information.
  • Double-Sided Formats Available
  • Three-Color Chart
  • Laminated Student Fill-In Form
  • Radioisotope Table
  • Subatomic Particle Table
Ward's comprehensive periodic table provides students with detailed and up-to-date information. In addition to elemental data, side one includes a table of radioisotopes with mass number, half-life, and decay particles. Side two includes a table of subatomic particles. These three-color charts come in a variety of sizes to meet all your teaching needs. Information updatedMore Product Information
Order Now
SPECIFICATIONS
Environmentally Preferable