Introduction to Reaction Rates
Supplier: Ward's Science
IS2512
470165-504EA 153.25 USD
470165-504
Introduction to Reaction Rates
Educational Materials Chemistry Educational Materials Mixtures and Solutions Learning Activities
How fast do chemical reactions happen?
  • Introduction to Concepts of Reaction Rates
  • Students Learn the Ways Different Factors Affect Speeds of Chemical Reactions
  • Activities Demonstrate How Scientists Adjust Reaction Rates by Understanding Factors that Influence Them
  • Experiments Explore Impacts of Concentration, Heat, Surface Area & Catalysts
  • Some Materials Included
Chemical reactions vary greatly. For example, some chemicals react extremely fast, like explosions, but others, like rusting metal, can react over a long period of time. Understanding theMore Product Information